Learn More
Avanti Polar Lipids 18:1 (Δ9-Cis) PE (DOPE), 4004-05-1, MFCD16621009, powder, 500mg
Catalog No. NC1681744
Encompass
Molecular Formula C41H78NO8P, Purity >99%, Formula Weight 744.034, Storage Temperature -20°C, 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine
Catalog No. NC1681744 Supplier Avanti Polar Lipids Supplier No. 850725P500MG
Molecular Formula C41H78NO8P, Purity >99%, Formula Weight 744.034, Storage Temperature -20°C, 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine