Learn More
Medchemexpress LLC 1,2-Dioleoyl-sn-glycero-3-phospho-L-serine (sodium) | 90693-88-2 | 98.0% | C42H77NNaO10P | 25 MG
SDP
Catalog No. 5000352914
Encompass_Preferred
1,2-Dioleoyl-sn-glycero-3-phospho-L-serine sodium (DOPS-NA) is a substitute for Phosphoserine/phosphatidylserine. It can be used with DOPC and DOPE in lipid mixtures for synthesizing liposomes. This compound can self-assemble into single-layer or double-layer membrane structures, resembling cell membranes, and exhibits high membrane fluidity and flexibility.
  • Substitute for phosphoserine/phosphatidylserine
  • Suitable for liposome synthesis with DOPC and DOPE
  • Self-assembles into single or double-layer membrane structures
  • Exhibits high membrane fluidity and flexibility
  • Widely used in membrane biology
  • Ideal for cell membrane research
  • Useful for lipid preparation
  • Applicable in drug delivery systems
Catalog No. 50-003-52914 Supplier Medchemexpress LLC Supplier No. 25MGHY141613
1,2-Dioleoyl-sn-glycero-3-phospho-L-serine sodium (DOPS-NA) is a substitute for Phosphoserine/phosphatidylserine. It can be used with DOPC and DOPE in lipid mixtures for synthesizing liposomes. This compound can self-assemble into single-layer or double-layer membrane structures, resembling cell membranes, and exhibits high membrane fluidity and flexibility.
  • Substitute for phosphoserine/phosphatidylserine
  • Suitable for liposome synthesis with DOPC and DOPE
  • Self-assembles into single or double-layer membrane structures
  • Exhibits high membrane fluidity and flexibility
  • Widely used in membrane biology
  • Ideal for cell membrane research
  • Useful for lipid preparation
  • Applicable in drug delivery systems