Learn More
Medchemexpress LLC 1,2-Dioleoyl-sn-glycero-3-phospho[N-(4-carboxybutanoyl)ethanolamine] | 228706-30-7 | C46H84NO11P | 100 MG
SDP
Catalog No. 5000375699
Encompass_Preferred
DOPE-GA can be used in the preparation of liposomes for research on drug delivery and other applications. It is one of the key raw materials for preparing liposome ELL-12 (a non-myelosuppressive antiproliferative agent with anti-tumor activity) and may affect the structure and stability of liposomes and the interaction between ELL-12 and cells.
  • Used in liposome preparation for drug delivery research
  • Key raw material for liposome ELL-12
  • Affects liposome structure and stability
  • Influences interaction between ELL-12 and cells
Catalog No. 50-003-75699 Supplier Medchemexpress LLC Supplier No. HY150240100MG
DOPE-GA can be used in the preparation of liposomes for research on drug delivery and other applications. It is one of the key raw materials for preparing liposome ELL-12 (a non-myelosuppressive antiproliferative agent with anti-tumor activity) and may affect the structure and stability of liposomes and the interaction between ELL-12 and cells.
  • Used in liposome preparation for drug delivery research
  • Key raw material for liposome ELL-12
  • Affects liposome structure and stability
  • Influences interaction between ELL-12 and cells