Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dioleoyl-sn-glycero-3-phosphocholine 97.0+%, TCI America™
SDP

Catalog No. d4250250mg Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4250250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4250250MG Supplier TCI America Supplier No. D4250250MG
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4235-95-4
Molecular Formula C44H84NO8P
Molecular Weight (g/mol) 786.13
MDL Number MFCD00135191
InChI Key SNKAWJBJQDLSFF-AOMFJDGTSA-N
Synonym DOPC, (R)-2,3-Bis(oleoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate
PubChem CID 10350317
ChEBI CHEBI:74669
IUPAC Name (2-{[(2S)-2,3-bis[(9Z)-octadec-9-enoyloxy]propyl phosphonato]oxy}ethyl)trimethylazanium
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OC[C@@H](COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCCCCC\C=C/CCCCCCCC

Specifications

Color White
Quantity 250 mg
Formula Weight 786.13
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Dioleoyl-sn-glycero-3-phosphocholine
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.