Learn More
Medchemexpress LLC 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[carboxy(polyethylene glycol)-2000] (sodium salt) | 2342575-87-3 | 95.0% | 2,000 Da (average) | C131H253NNaO55P (average, polydisperse PEG) | 25 MG
SDP
Catalog No. 5000281472
Encompass_Preferred
DOPE-PEG-COOH (MW 2000) is a PEGylated phospholipid composed of dioleoyl phosphoethanolamine linked to a carboxyl-terminated polyethylene glycol chain (average molecular weight 2,000 Da). It is supplied as a research-use reagent for use in liposome and nanoparticle formulation, surface functionalization, and bioconjugation/linker chemistry. Typical purity is 95.0% and the material is soluble in DMSO.
Catalog No. 50-002-81472 Supplier Medchemexpress LLC Supplier No. HYW44082725MG
DOPE-PEG-COOH (MW 2000) is a PEGylated phospholipid composed of dioleoyl phosphoethanolamine linked to a carboxyl-terminated polyethylene glycol chain (average molecular weight 2,000 Da). It is supplied as a research-use reagent for use in liposome and nanoparticle formulation, surface functionalization, and bioconjugation/linker chemistry. Typical purity is 95.0% and the material is soluble in DMSO.