Learn More
Medchemexpress LLC 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-2000] (ammonium salt) | 474922-90-2 | MFCD02262251 | 95.0% | 2,801.5 g/mol | (C2H4O)nC43H80NO10P.H3N | 25 MG
SDP
Catalog No. 5000281456
Encompass_Preferred
DOPE-mPEG (PEG 2000) is a PEGylated phospholipid designed for formulation of liposomes and giant unilamellar vesicles. The PEG surface layer improves circulation stability and reduces pH-sensitivity of liposomal membranes in a concentration-dependent manner, making it suitable for targeted delivery and surface functionalization of vesicles.
  • PEGylated phospholipid for liposome and GUV formulation.
  • Provides steric stabilization and reduced opsonization.
  • Reduces liposome pH-sensitivity in a concentration-dependent manner.
  • Average PEG chain molecular weight ~2000 Da.
  • Supplied as a solid, white to light yellow powder with ~95.0% purity.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • Suitable for targeted drug delivery and surface functionalization applications.
Catalog No. 50-002-81456 Supplier Medchemexpress LLC Supplier No. HYW44098825MG
DOPE-mPEG (PEG 2000) is a PEGylated phospholipid designed for formulation of liposomes and giant unilamellar vesicles. The PEG surface layer improves circulation stability and reduces pH-sensitivity of liposomal membranes in a concentration-dependent manner, making it suitable for targeted delivery and surface functionalization of vesicles.
  • PEGylated phospholipid for liposome and GUV formulation.
  • Provides steric stabilization and reduced opsonization.
  • Reduces liposome pH-sensitivity in a concentration-dependent manner.
  • Average PEG chain molecular weight ~2000 Da.
  • Supplied as a solid, white to light yellow powder with ~95.0% purity.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • Suitable for targeted drug delivery and surface functionalization applications.