Learn More
Medchemexpress LLC 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine N-(polyethylene glycol)-maleimide (sodium salt) | 98.2% | 2000 Da (average) | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW44099525MG
Encompass_Preferred
Maleimide-functionalized DOPE-PEG (PEG 2000) is a phospholipid-polyethylene glycol conjugate intended for the preparation of liposomes, micelles, and nanoparticles and for covalent attachment of thiol-containing ligands. The maleimide end-group reacts with thiols at pH 6.5-7.5 to form stable thioether bonds; material is supplied as a powder with an average PEG molecular weight of approximately 2000.
Catalog No. 50-002-81109