Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine 98.0+%, TCI America™
SDP

Catalog No. D4251250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4251250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4251250MG Supplier TCI America Supplier No. D4251250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4004-05-1
Molecular Formula C41H78NO8P
Molecular Weight (g/mol) 744.05
MDL Number MFCD00057986
InChI Key MWRBNPKJOOWZPW-NYVOMTAGSA-N
Synonym 2-Aminoethyl (R)-2,3-Bis(oleoyloxy)propyl Hydrogen Phosphate, DOPE
PubChem CID 54095275
IUPAC Name [(2R)-3-[2-aminoethoxy(hydroxy)phosphoryl]oxy-2-octadec-9-enoyloxypropyl] octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OCC(COP(=O)(O)OCCN)OC(=O)CCCCCCCC=CCCCCCCCC

Specifications

Melting Point 200°C
Color White-Yellow
Quantity 250 mg
Formula Weight 744.05
Percent Purity ≥98.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material 1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.