Learn More
Cayman Chemical 1,2-Dioleoyl-sn-glycerol, Purity: ≥95%, CAS: 24529-88-2, 100 mg
SDP
Catalog No. 502893559
Encompass_Preferred
An analog of the PKC-activating second messengers collectively called DAG; equipotent to 1,2-dioctanoyl-sn-glycerol in induction of the acrosome reaction in human sperm
Catalog No. 50-289-3559 Supplier Cayman Chemical Supplier No. 62230100 MG
An analog of the PKC-activating second messengers collectively called DAG; equipotent to 1,2-dioctanoyl-sn-glycerol in induction of the acrosome reaction in human sperm