Learn More
Medchemexpress LLC 1,2-Dioleoyl-sn-glycerol | 24529-88-2 | 100.0% | C39H72O5 | 50 MG
SDP
Catalog No. 5000356773
Encompass_Preferred
1,2-Dioleoyl-sn-glycerol is a glycerolipid found on the plasma membrane. It comprises two fatty acid chains linked by an ester bond to a single glycerol molecule. This compound acts as an analog of diacylglycerol (DAG), a second messenger that activates protein kinase C. It has been utilized as a diacylglycerol source in the diacylglycerol O-acyltransferase 1 (DGAT1) assay and can induce acrosome reaction in human sperm.
  • Glycerolipid found on the plasma membrane
  • Analog of diacylglycerol (DAG)
  • Activates protein kinase C
  • Used as a diacylglycerol source in DGAT1 assay
  • Induces acrosome reaction in human sperm
  • Intended for research use only
Catalog No. 50-003-56773 Supplier Medchemexpress LLC Supplier No. HY14157250MG
1,2-Dioleoyl-sn-glycerol is a glycerolipid found on the plasma membrane. It comprises two fatty acid chains linked by an ester bond to a single glycerol molecule. This compound acts as an analog of diacylglycerol (DAG), a second messenger that activates protein kinase C. It has been utilized as a diacylglycerol source in the diacylglycerol O-acyltransferase 1 (DGAT1) assay and can induce acrosome reaction in human sperm.
  • Glycerolipid found on the plasma membrane
  • Analog of diacylglycerol (DAG)
  • Activates protein kinase C
  • Used as a diacylglycerol source in DGAT1 assay
  • Induces acrosome reaction in human sperm
  • Intended for research use only