Learn More
Medchemexpress LLC 1,2-Dioleoyl-sn-glycerol | 24529-88-2 | >99.9% | C₃₉H₇₂O₅ | 25 MG
SDP
Catalog No. 5000352909
Encompass_Preferred
1,2-Dioleoyl-sn-glycerol is a glycerolipid found on the plasma membrane, composed of two fatty acid chains linked to a glycerol molecule via an ester bond. It functions as an analog of diacylglycerol (DAG), a protein kinase C-activating second messenger. This compound has been utilized as a diacylglycerol source in the diacylglycerol O-acyltransferase 1 (DGAT1) assay and can induce acrosome reaction in human sperm. It has a molecular weight of 620.99 and is considered a drug derivative. This product is for research use only.
  • Consists of two fatty acid chains covalently linked to a single glycerol molecule
  • Functions as an analog of protein kinase C-activating second messenger DAG
  • Used as a diacylglycerol source in DGAT1 assay
  • Can induce acrosome reaction in human sperm
  • Soluble in DMSO and ethanol
Catalog No. 50-003-52909 Supplier Medchemexpress LLC Supplier No. 25MGHY141572
1,2-Dioleoyl-sn-glycerol is a glycerolipid found on the plasma membrane, composed of two fatty acid chains linked to a glycerol molecule via an ester bond. It functions as an analog of diacylglycerol (DAG), a protein kinase C-activating second messenger. This compound has been utilized as a diacylglycerol source in the diacylglycerol O-acyltransferase 1 (DGAT1) assay and can induce acrosome reaction in human sperm. It has a molecular weight of 620.99 and is considered a drug derivative. This product is for research use only.
  • Consists of two fatty acid chains covalently linked to a single glycerol molecule
  • Functions as an analog of protein kinase C-activating second messenger DAG
  • Used as a diacylglycerol source in DGAT1 assay
  • Can induce acrosome reaction in human sperm
  • Soluble in DMSO and ethanol