Learn More
Medchemexpress LLC 1,2-dioleoyl-sn-glycerol | 24529-88-2 | MFCD00148866 | 100.0% | C39H72O5 | 10MG
SDP
Catalog No. 5000259420
Encompass_Preferred
1,2-Dioleoyl-sn-glycerol is a diacylglycerol lipid used as a biochemical research reagent and assay substrate. It provides a physiologically relevant 18:1/18:1 diacylglycerol species for enzyme and signaling studies, and is supplied with documentation for quality and safety. Intended for in vitro research use; follow storage and solvent recommendations for stability and solubility.
  • High purity suitable for biochemical assays.
  • Provides physiologically relevant 18:1/18:1 diacylglycerol species.
  • Supplied with COA and SDS for quality and safety documentation.
  • Soluble in DMSO and ethanol with ultrasonic assistance.
  • Stable when stored at -20°C; in solvent store at -80°C for extended periods.
  • Liquid form simplifies preparation of assay solutions.
Catalog No. 50-002-59420 Supplier Medchemexpress LLC Supplier No. HY14157210MG
1,2-Dioleoyl-sn-glycerol is a diacylglycerol lipid used as a biochemical research reagent and assay substrate. It provides a physiologically relevant 18:1/18:1 diacylglycerol species for enzyme and signaling studies, and is supplied with documentation for quality and safety. Intended for in vitro research use; follow storage and solvent recommendations for stability and solubility.
  • High purity suitable for biochemical assays.
  • Provides physiologically relevant 18:1/18:1 diacylglycerol species.
  • Supplied with COA and SDS for quality and safety documentation.
  • Soluble in DMSO and ethanol with ultrasonic assistance.
  • Stable when stored at -20°C; in solvent store at -80°C for extended periods.
  • Liquid form simplifies preparation of assay solutions.