Learn More
Cayman Chemical 1 2-DIoleoyl-sn-glycerol 25mg
SDP
Catalog No. 502892778
Encompass_Preferred
An analog of the PKC-activating second messengers collectively called DAG; equipotent to 1,2-dioctanoyl-sn-glycerol in induction of the acrosome reaction in human sperm
Catalog No. 50-289-2778 Supplier Cayman Chemical Supplier No. 6223025 MG
An analog of the PKC-activating second messengers collectively called DAG; equipotent to 1,2-dioctanoyl-sn-glycerol in induction of the acrosome reaction in human sperm