Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2-Dipalmitoyl-sn-glycero-3-phospho-N,N-dimethylethanolamine | 3922-61-0 | 99.0% | C39H78NO8P | 50 MG
SDP

Catalog No. 5000354168
Encompass_Preferred

1,2-Dipalmitoyl-sn-glycero-3-phospho-N,N-dimethylethanolamine is a white to off-white solid used in the generation of liposomes and monolayers. It is suitable for studying membrane permeability and monolayer viscosity. This product is intended for research use only.

  • Used in the generation of liposomes
  • Used in the generation of monolayers
  • Suitable for studying membrane permeability
  • Suitable for studying monolayer viscosity
  • Soluble in ethanol and H₂O at 2 mg/mL (2.78 mM) with ultrasonic, warming, and heating to 60 °C
  • Insoluble or slightly soluble in DMSO at less than 1 mg/mL
  • Store at room temperature, dry and cool
  • For storage in solvent, store at -80 °C for 1 year or -20 °C for 6 months

Catalog No. 50-003-54168 Supplier Medchemexpress LLC Supplier No. HY14161550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,2-Dipalmitoyl-sn-glycero-3-phospho-N,N-dimethylethanolamine is a white to off-white solid used in the generation of liposomes and monolayers. It is suitable for studying membrane permeability and monolayer viscosity. This product is intended for research use only.

  • Used in the generation of liposomes
  • Used in the generation of monolayers
  • Suitable for studying membrane permeability
  • Suitable for studying monolayer viscosity
  • Soluble in ethanol and H₂O at 2 mg/mL (2.78 mM) with ultrasonic, warming, and heating to 60 °C
  • Insoluble or slightly soluble in DMSO at less than 1 mg/mL
  • Store at room temperature, dry and cool
  • For storage in solvent, store at -80 °C for 1 year or -20 °C for 6 months

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.