Learn More
Medchemexpress LLC 1,2-dipalmitoyl-sn-glycero-3-phospho-N,N-dimethylethanolamine | 3922-61-0 | MFCD00066514 | 99.0% | C39H78NO8P | 10MG
SDP
Supplier:  Medchemexpress LLC HY14161510MG
Encompass_Preferred
1,2-Dipalmitoyl-sn-glycero-3-phospho-N,N-dimethylethanolamine is a synthetic dimethyl phosphoethanolamine phospholipid with two palmitoyl (16:0) acyl chains. Supplied as a high-purity solid, it is intended for liposome formulation and membrane biophysics research, providing consistent physical and chemical properties for experimental reproducibility.
  • High purity (99.0%) for reproducible results.
  • Palmitoyl (16:0) acyl chains produce saturated lipid behavior.
  • Solid, white to off-white appearance for easy handling and storage.
  • Suitable for liposome preparation and membrane permeability studies.
  • Available in small quantities to support formulation optimization.
  • Certificate of analysis and safety data sheet available for quality and safety documentation.
Catalog No. 50-002-61545