Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine 96.0+%, TCI America™
SDP

Catalog No. D4213250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4213250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4213250MG Supplier TCI America Supplier No. D4213250MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 923-61-5
Molecular Formula C37H74NO8P
Molecular Weight (g/mol) 691.972
MDL Number MFCD00036776
InChI Key SLKDGVPOSSLUAI-PGUFJCEWSA-N
Synonym 1,2-Dihexadecanoyl-sn-glycero-3-phosphoethanolamine, Dipalmitoylphosphatidylethanolamine, 2-Aminoethyl (R)-2,3-Bis(hexadecanoyloxy)propyl Hydrogen Phosphate, L-DPPE
PubChem CID 445468
ChEBI CHEBI:73127
IUPAC Name [(2R)-3-[2-aminoethoxy(hydroxy)phosphoryl]oxy-2-hexadecanoyloxypropyl] hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)(O)OCCN)OC(=O)CCCCCCCCCCCCCCC

Specifications

Melting Point 197°C
Color White
Quantity 250 mg
Formula Weight 691.97
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.