Learn More
Medchemexpress LLC 1,2-dipalmitoyl-sn-glycero-3-phosphoglycerol sodium | 200880-41-7 | 98.0% | 744.95 g/mol | C38H74NaO10P | 250MG
SDP
Catalog No. 5000235319
Encompass_Preferred
1,2-Dipalmitoyl-sn-glycero-3-phosphoglycerol sodium is a long-chain anionic phospholipid composed of two palmitoyl (16:0) fatty acyl chains on a glycerol backbone with a phosphoglycerol head group. It is commonly used in membrane model systems, liposome formulation, and biophysical studies of phospholipid bilayers due to its defined phase behavior and reproducible properties.
  • High purity (98.0%) suitable for research applications.
  • Anionic phospholipid for liposome and membrane model preparation.
  • Well-defined molecular weight and formula for reproducible experiments.
  • Available in multiple pack sizes for small-scale and bulk studies.
  • Solid, white to off-white appearance for easy handling and formulation.
Catalog No. 50-002-35319 Supplier Medchemexpress LLC Supplier No. HY125940250MG
1,2-Dipalmitoyl-sn-glycero-3-phosphoglycerol sodium is a long-chain anionic phospholipid composed of two palmitoyl (16:0) fatty acyl chains on a glycerol backbone with a phosphoglycerol head group. It is commonly used in membrane model systems, liposome formulation, and biophysical studies of phospholipid bilayers due to its defined phase behavior and reproducible properties.
  • High purity (98.0%) suitable for research applications.
  • Anionic phospholipid for liposome and membrane model preparation.
  • Well-defined molecular weight and formula for reproducible experiments.
  • Available in multiple pack sizes for small-scale and bulk studies.
  • Solid, white to off-white appearance for easy handling and formulation.