Learn More
Medchemexpress LLC 1,2-Dipalmitoyl-sn-glycerol 3-phosphate sodium | 169051-60-9 | 98.0% | 670.87 | 25 MG
SDP
Catalog No. 5000438405
Encompass_Preferred
1,2-Dipalmitoyl-sn-glycerol 3-phosphate sodium (compound 3-F7) is identified as a phosphatidic acid and a human endogenous metabolite. It is utilized in the creation of micelles, liposomes, and artificial membranes. Phosphatidic acids are biologically active lipids that can stimulate various responses in different cell types, including platelet aggregation, smooth muscle contraction, in vivo vasoactive effects, chemotaxis, expression of adhesion molecules, increased tight junction permeability of endothelial cells, induction of stress fibers, and modulation of cardiac contractility.
  • Human endogenous metabolite
  • Used in the generation of micelles, liposomes, and artificial membranes
  • Biologically active lipid
Catalog No. 50-004-38405 Supplier Medchemexpress LLC Supplier No. 25MGHY113437A
1,2-Dipalmitoyl-sn-glycerol 3-phosphate sodium (compound 3-F7) is identified as a phosphatidic acid and a human endogenous metabolite. It is utilized in the creation of micelles, liposomes, and artificial membranes. Phosphatidic acids are biologically active lipids that can stimulate various responses in different cell types, including platelet aggregation, smooth muscle contraction, in vivo vasoactive effects, chemotaxis, expression of adhesion molecules, increased tight junction permeability of endothelial cells, induction of stress fibers, and modulation of cardiac contractility.
  • Human endogenous metabolite
  • Used in the generation of micelles, liposomes, and artificial membranes
  • Biologically active lipid