Learn More
Medchemexpress LLC 1,2-Dipalmitoyl-sn-glycerol 3-phosphate sodium | 169051-60-9 | 98.0% | 670.87 | 50 MG
Supplier: Medchemexpress LLC HY113437A50MG
1,2-Dipalmitoyl-sn-glycerol 3-phosphate sodium (compound 3-F7) is a phosphatidic acid and a human endogenous metabolite. It is used in the generation of micelles, liposomes, and artificial membranes. Phosphatidic acids are biologically active lipids that can stimulate a large range of responses in many different cell types, for research use only.
- Stimulates platelet aggregation
- Induces smooth muscle contraction
- Exhibits in vivo vasoactive effects
- Supports chemotaxis
- Increases tight junction permeability of endothelial cells
- Induces stress fibres
- Modulates cardiac contractility