Learn More
Medchemexpress LLC 1,2-dipalmitoyl-sn-glycerol 3-phosphate sodium | 169051-60-9 | 98.0% | 670.87 g/mol | C35H68NaO8P | 5 MG
SDP
Supplier:  Medchemexpress LLC HY113437A5MG
Encompass_Preferred
1,2-Dipalmitoyl-sn-glycerol 3-phosphate sodium is a phosphatidic acid lipid provided as the sodium salt for use in biochemical research and lipidomics. It is supplied as a solid with a reported purity of 98.0%, molecular weight 670.87 g/mol, and formula C35H68NaO8P. SDS and certificate of analysis are available to support safe handling and quality control.
  • High purity (98.0%) suitable for analytical and preparative work.
  • Solid form for stable handling and storage.
  • Validated molecular weight and formula for identification and QC.
  • Multiple small pack sizes for experimental flexibility.
  • Access to SDS and certificate of analysis for regulatory compliance.
  • Recommended storage: -20°C sealed; in solvent: -80°C for long-term.
Catalog No. 50-002-64803