Learn More
Medchemexpress LLC 1,2-Dipalmitoyl-sn-glycerol | 30334-71-5 | 98.0% | 568.92 | 25 MG
SDP
Catalog No. 5000451421
Encompass_Preferred
1,2-Dipalmitoyl-sn-glycerol is an endogenous metabolite for research use only. It is a solid, white to off-white in appearance, commonly used in laboratory settings.
  • Endogenous metabolite
  • Supplied as a solid
  • Appearance: white to off-white powder
  • Store powder at -20°C for up to 3 years
Catalog No. 50-004-51421 Supplier Medchemexpress LLC Supplier No. 25MGHYW010736
1,2-Dipalmitoyl-sn-glycerol is an endogenous metabolite for research use only. It is a solid, white to off-white in appearance, commonly used in laboratory settings.
  • Endogenous metabolite
  • Supplied as a solid
  • Appearance: white to off-white powder
  • Store powder at -20°C for up to 3 years