Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2-Diphenylethane-d14 | 94371-89-8 | 99.5% | C14D14 | 100 MG
SDP

Catalog No. 5000433194
Encompass_Preferred

1,2-Diphenylethane-d14 is the deuterium labeled 1,2-Diphenylethane. It is intended for research use only.

  • Used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration impacts pharmacokinetic and metabolic profiles of drugs
  • Molecular weight of 196.35
  • Appearance as a crystal
  • Color is white to off-white
  • Storage conditions: powder -20°C for 3 years; in solvent -80°C for 6 months and -20°C for 1 month

Catalog No. 50-004-33194 Supplier Medchemexpress LLC Supplier No. HY78004S100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,2-Diphenylethane-d14 is the deuterium labeled 1,2-Diphenylethane. It is intended for research use only.

  • Used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration impacts pharmacokinetic and metabolic profiles of drugs
  • Molecular weight of 196.35
  • Appearance as a crystal
  • Color is white to off-white
  • Storage conditions: powder -20°C for 3 years; in solvent -80°C for 6 months and -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.