Learn More
Medchemexpress LLC 1,2-Diphenylethane-d14 | 94371-89-8 | 99.5% | C14D14 | 25 MG
SDP
Catalog No. 5000358265
Encompass_Preferred
1,2-Diphenylethane-d14 is a deuterium-labeled form of 1,2-Diphenylethane. It is intended for research use only and serves as a valuable tool in quantitative analysis. Deuteration studies are significant for their potential impact on drug pharmacokinetic and metabolic profiles. This compound has a molecular weight of 196.35 and appears as a white to off-white crystal.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Applicable in techniques such as NMR, GC-MS, or LC-MS
Catalog No. 50-003-58265 Supplier Medchemexpress LLC Supplier No. 25MGHY78004S
1,2-Diphenylethane-d14 is a deuterium-labeled form of 1,2-Diphenylethane. It is intended for research use only and serves as a valuable tool in quantitative analysis. Deuteration studies are significant for their potential impact on drug pharmacokinetic and metabolic profiles. This compound has a molecular weight of 196.35 and appears as a white to off-white crystal.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Applicable in techniques such as NMR, GC-MS, or LC-MS