Learn More
Medchemexpress LLC 1,2-Diphenylethane-d14 | 94371-89-8 | 99.5% | C14D14 | 250 MG
SDP
Catalog No. 5000358624
Encompass_Preferred
1,2-Diphenylethane-d14 is the deuterium labeled 1,2-Diphenylethane, intended for research use. This compound can be utilized as a tracer and as an internal standard for quantitative analysis through techniques such as Nuclear Magnetic Resonance (NMR), Gas Chromatography-Mass Spectrometry (GC-MS), or Liquid Chromatography-Mass Spectrometry (LC-MS). The deuteration of drug molecules has garnered attention due to its potential to influence pharmacokinetic and metabolic profiles of drugs.
  • Intended for research use
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, or LC-MS analytical methods
  • Deuteration can affect pharmacokinetic and metabolic profiles of drug molecules
Catalog No. 50-003-58624 Supplier Medchemexpress LLC Supplier No. 250MGHY78004S
1,2-Diphenylethane-d14 is the deuterium labeled 1,2-Diphenylethane, intended for research use. This compound can be utilized as a tracer and as an internal standard for quantitative analysis through techniques such as Nuclear Magnetic Resonance (NMR), Gas Chromatography-Mass Spectrometry (GC-MS), or Liquid Chromatography-Mass Spectrometry (LC-MS). The deuteration of drug molecules has garnered attention due to its potential to influence pharmacokinetic and metabolic profiles of drugs.
  • Intended for research use
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, or LC-MS analytical methods
  • Deuteration can affect pharmacokinetic and metabolic profiles of drug molecules