Learn More
Medchemexpress LLC 1,2-Diphenylethane-d14 | 94371-89-8 | 99.5% | C14D14 | 50 MG
SDP
Catalog No. 5000432585
Encompass_Preferred
1,2-Diphenylethane-d14 is the deuterium labeled 1,2-Diphenylethane. It is for research use only and not sold to patients. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during drug development. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-32585 Supplier Medchemexpress LLC Supplier No. HY78004S50MG
1,2-Diphenylethane-d14 is the deuterium labeled 1,2-Diphenylethane. It is for research use only and not sold to patients. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during drug development. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.