Learn More
Medchemexpress LLC 1-(2-(((diphenylmethylene)amino)oxy)ethyl)-1,2,5,6-tetrahydro-3-pyridinecarboxylic acid hydrochloride | 145645-62-1 | MFCD00153853 | 99.8% | 386.87 g/mol | C21H23ClN2O3 | 25 MG
SDP
Catalog No. 5000248626
Encompass_Preferred
NNC-711 (NO-711 hydrochloride) is a potent and selective inhibitor of the GABA transporter GAT-1 supplied as a high-purity solid for research applications in neuroscience and pharmacology. It exhibits anticonvulsant and analgesic activity in vivo and is used to study GABA uptake and transporter function.
  • Potent and selective GAT-1 inhibitor; IC50 ≈ 0.04 μM for human GAT-1.
  • High purity suitable for in vitro and in vivo research.
  • White to off-white solid, supplied in small pack sizes for laboratory use.
  • Stable when stored sealed at low temperature, away from moisture.
  • For research use only; not for human or clinical applications.
Catalog No. 50-002-48626 Supplier Medchemexpress LLC Supplier No. HY10350625MG
NNC-711 (NO-711 hydrochloride) is a potent and selective inhibitor of the GABA transporter GAT-1 supplied as a high-purity solid for research applications in neuroscience and pharmacology. It exhibits anticonvulsant and analgesic activity in vivo and is used to study GABA uptake and transporter function.
  • Potent and selective GAT-1 inhibitor; IC50 ≈ 0.04 μM for human GAT-1.
  • High purity suitable for in vitro and in vivo research.
  • White to off-white solid, supplied in small pack sizes for laboratory use.
  • Stable when stored sealed at low temperature, away from moisture.
  • For research use only; not for human or clinical applications.