Learn More
Medchemexpress LLC 1,2-distearoyl-3-chloropropanediol (racemic) | 72468-92-9 | MFCD18452167 | >99.0% | 643.46 g·mol⁻¹ | C39H75ClO4 | 10 MG
SDP
Catalog No. 5000282647
Encompass_Preferred
rac-1,2-distearoyl-3-chloropropanediol is a racemic distearoyl ester of 3-chloropropanediol used as a lipid-class biochemical reagent and analytical reference material for life-science research. It is suitable for method development, analytical testing, and studies that require defined fatty acyl esters and reproducible lipid standards.
  • Lipid-class biochemical reagent suitable for analytical and research applications.
  • Racemic form for studies requiring ± isomer composition.
  • High purity >99.0% for reproducible analytical results.
  • CAS number 72468-92-9 for unambiguous identification.
  • Supplied in small pack sizes for method development and testing.
Catalog No. 50-002-82647 Supplier Medchemexpress LLC Supplier No. HYW12757010MG
Add to Cart
Add to Cart
rac-1,2-distearoyl-3-chloropropanediol is a racemic distearoyl ester of 3-chloropropanediol used as a lipid-class biochemical reagent and analytical reference material for life-science research. It is suitable for method development, analytical testing, and studies that require defined fatty acyl esters and reproducible lipid standards.
  • Lipid-class biochemical reagent suitable for analytical and research applications.
  • Racemic form for studies requiring ± isomer composition.
  • High purity >99.0% for reproducible analytical results.
  • CAS number 72468-92-9 for unambiguous identification.
  • Supplied in small pack sizes for method development and testing.