Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2-distearoyl-d70-sn-glycero-3-phosphocholine | 56952-01-3 | MFCD00674391 | 99.0% | 860.577 | C44H18D70NO8P | 5 MG
SDP

Catalog No. 5000208787
Encompass_Preferred

DSPC-d70 is the deuterium-labeled form of 1,2-distearoyl-sn-glycero-3-phosphocholine used as an analytical tracer and as a lipid component in liposome and lipid nanoparticle formulations.

  • Perdeuterated phosphatidylcholine (d70).
  • Used as a tracer or internal standard for NMR, GC-MS, and LC-MS.
  • Suitable for liposome and lipid nanoparticle formulation.
  • Purity 99.0%.
  • CAS number 56952-01-3.
  • Molecular weight 860.577.
  • Packaged as 5 MG.

Catalog No. 50-002-08787 Supplier Medchemexpress LLC Supplier No. HYW040193S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DSPC-d70 is the deuterium-labeled form of 1,2-distearoyl-sn-glycero-3-phosphocholine used as an analytical tracer and as a lipid component in liposome and lipid nanoparticle formulations.

  • Perdeuterated phosphatidylcholine (d70).
  • Used as a tracer or internal standard for NMR, GC-MS, and LC-MS.
  • Suitable for liposome and lipid nanoparticle formulation.
  • Purity 99.0%.
  • CAS number 56952-01-3.
  • Molecular weight 860.577.
  • Packaged as 5 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.