Learn More
Medchemexpress LLC 1 2-Distearoyl-sn-gl 100mg | 108321-18-2 | 100 MG
SDP
Catalog No. 5000227096
Encompass_Preferred
1,2-Distearoyl-sn-glycero-3-phosphate sodium is a phospholipid (sodium salt) used in research for liposome formulation and drug delivery studies. It acts as a stabilizer and emulsifier to aid in drug encapsulation and improve solubility and bioavailability. Store sealed and protected from moisture for best stability.
  • phospholipid useful for liposome formulations
  • functions as a stabilizer and emulsifier
  • solid, white to off-white appearance
  • available in small laboratory pack sizes for research use
  • recommended sealed storage to minimize moisture exposure
Catalog No. 50-002-27096 Supplier Medchemexpress LLC Supplier No. HY115415100MG
1,2-Distearoyl-sn-glycero-3-phosphate sodium is a phospholipid (sodium salt) used in research for liposome formulation and drug delivery studies. It acts as a stabilizer and emulsifier to aid in drug encapsulation and improve solubility and bioavailability. Store sealed and protected from moisture for best stability.
  • phospholipid useful for liposome formulations
  • functions as a stabilizer and emulsifier
  • solid, white to off-white appearance
  • available in small laboratory pack sizes for research use
  • recommended sealed storage to minimize moisture exposure