Learn More
Medchemexpress LLC 1 2-Distearoyl-sn-gl 25mg | 108321-18-2 | 726.98 | C39H76NaO8P | 25 MG
SDP
Catalog No. 5000308016
Encompass_Preferred
1,2-Distearoyl-sn-glycero-3-phosphate sodium is the sodium salt of a distearoyl phosphatidic acid (C18:0 phospholipid). Supplied as a solid, high-purity reagent, it is used in lipid formulations, liposome and lipid nanoparticle preparation, and biochemical assays.
  • High purity suitable for research applications.
  • Solid form for straightforward handling and storage.
  • Soluble in methanol with ultrasonic warming (approx. 2 mg/mL).
  • Intended for liposome and lipid nanoparticle assembly and lipid formulation studies.
  • Available in small pack sizes appropriate for lab-scale experiments.
Catalog No. 50-003-08016 Supplier Medchemexpress LLC Supplier No. HY11541525MG
1,2-Distearoyl-sn-glycero-3-phosphate sodium is the sodium salt of a distearoyl phosphatidic acid (C18:0 phospholipid). Supplied as a solid, high-purity reagent, it is used in lipid formulations, liposome and lipid nanoparticle preparation, and biochemical assays.
  • High purity suitable for research applications.
  • Solid form for straightforward handling and storage.
  • Soluble in methanol with ultrasonic warming (approx. 2 mg/mL).
  • Intended for liposome and lipid nanoparticle assembly and lipid formulation studies.
  • Available in small pack sizes appropriate for lab-scale experiments.