Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1 2-Distearoyl-sn-gl 50mg | 108321-18-2 | 50 MG
SDP

Catalog No. 5000228259
Encompass_Preferred

1,2-distearoyl-sn-glycero-3-phosphate sodium is a phospholipid (sodium salt) used to formulate liposomes and lipid nanoparticles, providing structural stability and a negatively charged phosphate headgroup for drug delivery research.

  • Phospholipid with two C18 stearoyl acyl chains.
  • Provides negative surface charge for lipid assemblies.
  • Molecular weight 726.98 g·mol⁻¹ and formula C39H76NaO8P.
  • High purity (98.0%) white to off-white solid.
  • Soluble in methanol with warming; poorly soluble in DMSO.
  • Supplied in small research pack sizes and suitable for formulation studies.

Catalog No. 50-002-28259 Supplier Medchemexpress LLC Supplier No. HY11541550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,2-distearoyl-sn-glycero-3-phosphate sodium is a phospholipid (sodium salt) used to formulate liposomes and lipid nanoparticles, providing structural stability and a negatively charged phosphate headgroup for drug delivery research.

  • Phospholipid with two C18 stearoyl acyl chains.
  • Provides negative surface charge for lipid assemblies.
  • Molecular weight 726.98 g·mol⁻¹ and formula C39H76NaO8P.
  • High purity (98.0%) white to off-white solid.
  • Soluble in methanol with warming; poorly soluble in DMSO.
  • Supplied in small research pack sizes and suitable for formulation studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.