Learn More
Medchemexpress LLC HY-W040193 500mg , 1,2-Distearoyl-sn-glycero-3-phosphorylcholine CAS:816-94-4 Purity:>98%
SDP
Catalog No. 502261600
Encompass_Preferred
HY-W040193 500mg 1,2-Distearoyl-sn-glycero-3-phosphorylcholine CAS: 816-94-4 1,2-Distearoyl-sn-glycero-3-phosphorylcholine (1,2-Distearoyl-sn-glycero-3-PC; DSPC) is a cylindrical-shaped lipid. 1,2-Distearoyl-sn-glycero-3-phosphorylcholine is used to synthesize liposomes, and is the lipid component in the lipid nanoparticle (LNP) system. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1600 Supplier Medchemexpress LLC Supplier No. HYW040193500MG
HY-W040193 500mg 1,2-Distearoyl-sn-glycero-3-phosphorylcholine CAS: 816-94-4 1,2-Distearoyl-sn-glycero-3-phosphorylcholine (1,2-Distearoyl-sn-glycero-3-PC; DSPC) is a cylindrical-shaped lipid. 1,2-Distearoyl-sn-glycero-3-phosphorylcholine is used to synthesize liposomes, and is the lipid component in the lipid nanoparticle (LNP) system. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.