Learn More
TARGETMOL CHEMICALS INC 1 2-Distearoyl-sn-glycero-3-ph
SDP
Catalog No. 502879124
Encompass_Preferred
Also available in 100 mg, 200 mg and bulk. Please contact Fisher for quotes. 1,2-Distearoyl-sn-glycero-3-phosphorylcholine (L-ALPHA-PHOSPHATIDYLCHOLINE, DISTEAROYL) is a cylindrical-shaped lipid used to synthesize liposomes. 1,2-Distearoyl-sn-glycero-3-phosphorylcholine (L-ALPHA-PHOSPHATIDYLCHOLINE, DISTEAROYL) is the lipid component in the lipid nanoparticle (LNP) system. Purity 99.53%
Catalog No. 50-287-9124 Supplier TARGETMOL CHEMICALS INC Supplier No. T9510500MG
Also available in 100 mg, 200 mg and bulk. Please contact Fisher for quotes. 1,2-Distearoyl-sn-glycero-3-phosphorylcholine (L-ALPHA-PHOSPHATIDYLCHOLINE, DISTEAROYL) is a cylindrical-shaped lipid used to synthesize liposomes. 1,2-Distearoyl-sn-glycero-3-phosphorylcholine (L-ALPHA-PHOSPHATIDYLCHOLINE, DISTEAROYL) is the lipid component in the lipid nanoparticle (LNP) system. Purity 99.53%