Learn More
Medchemexpress LLC 1,2-Distearoyl-sn-Glycero-3-Phosphatidylglycerol sodium | 200880-42-8 | MFCD00077290 | 98.0% | C42H82NaO10P | 250 MG
SDP
Catalog No. 5000355930
Encompass_Preferred
1,2-Distearoyl-sn-Glycero-3-Phosphatidylglycerol sodium is a phospholipid compound primarily used as a component in liposomes for drug delivery systems. It is instrumental in preparing negatively charged nanoliposomes, which have demonstrated efficacy in correcting dyslipidemia and inhibiting atherosclerosis progression in high-cholesterol diet models.
  • Component of liposomes for drug delivery
  • Can prepare negatively charged nanoliposomes
  • Used in research for blood or cardio-cerebrovascular disease
  • Relevant for cardiovascular disease and metabolic enzyme/protease liposome studies
Catalog No. 50-003-55930 Supplier Medchemexpress LLC Supplier No. 250MGHY145742
1,2-Distearoyl-sn-Glycero-3-Phosphatidylglycerol sodium is a phospholipid compound primarily used as a component in liposomes for drug delivery systems. It is instrumental in preparing negatively charged nanoliposomes, which have demonstrated efficacy in correcting dyslipidemia and inhibiting atherosclerosis progression in high-cholesterol diet models.
  • Component of liposomes for drug delivery
  • Can prepare negatively charged nanoliposomes
  • Used in research for blood or cardio-cerebrovascular disease
  • Relevant for cardiovascular disease and metabolic enzyme/protease liposome studies