Learn More
Medchemexpress LLC 1,2-Distearoyl-sn-Glycero-3-Phosphatidylglycerol sodium | 200880-42-8 | MFCD00077290 | 98.0% | C42H82NaO10P | 50 MG
SDP
Catalog No. 5000355941
Encompass_Preferred
1,2-Distearoyl-sn-Glycero-3-Phosphatidylglycerol (sodium) is a component of liposomes used for drug delivery. It can be used to prepare negatively charged nanoliposomes. These nanoliposome formulations have been shown to correct dyslipidaemia and inhibit the progression of atherosclerosis in rabbits fed a high cholesterol diet.
  • Component of liposomes for drug delivery
  • Used to prepare negatively charged nanoliposomes
  • Nanoliposome formulations correct dyslipidaemia
  • Nanoliposome formulations inhibit atherosclerosis progression
  • For research use only
Catalog No. 50-003-55941 Supplier Medchemexpress LLC Supplier No. HY14574250MG
1,2-Distearoyl-sn-Glycero-3-Phosphatidylglycerol (sodium) is a component of liposomes used for drug delivery. It can be used to prepare negatively charged nanoliposomes. These nanoliposome formulations have been shown to correct dyslipidaemia and inhibit the progression of atherosclerosis in rabbits fed a high cholesterol diet.
  • Component of liposomes for drug delivery
  • Used to prepare negatively charged nanoliposomes
  • Nanoliposome formulations correct dyslipidaemia
  • Nanoliposome formulations inhibit atherosclerosis progression
  • For research use only