Learn More
Medchemexpress LLC 1,2-distearoyl-sn-glycero-3-phosphocholine-d70 | 56952-01-3 | >99.0% | 860.58 g/mol | C44H18D70NO8P | 1 MG
SDP
Catalog No. 5000209730
Encompass_Preferred
DSPC-d70 (1,2-distearoyl-sn-glycero-3-phosphocholine-d70) is the deuterium-labeled form of a fully saturated phospholipid used in liposome and lipid nanoparticle research and analytical isotope-labeling studies. The d70 isotopologue replaces hydrogen with deuterium to enable improved detection in mass spectrometry and NMR, and to act as an isotopic tracer in formulation stability and biodistribution experiments.
  • Deuterium-labeled phospholipid for isotope tracing and analytical studies.
  • Suitable for liposome and lipid nanoparticle formulations.
  • High molecular weight and full saturation for stable bilayer formation.
  • Useful in mass spectrometry and NMR applications to simplify spectral interpretation.
  • Available in small pack sizes for research-scale experiments.
Catalog No. 50-002-09730 Supplier Medchemexpress LLC Supplier No. HYW040193S1MG
DSPC-d70 (1,2-distearoyl-sn-glycero-3-phosphocholine-d70) is the deuterium-labeled form of a fully saturated phospholipid used in liposome and lipid nanoparticle research and analytical isotope-labeling studies. The d70 isotopologue replaces hydrogen with deuterium to enable improved detection in mass spectrometry and NMR, and to act as an isotopic tracer in formulation stability and biodistribution experiments.
  • Deuterium-labeled phospholipid for isotope tracing and analytical studies.
  • Suitable for liposome and lipid nanoparticle formulations.
  • High molecular weight and full saturation for stable bilayer formation.
  • Useful in mass spectrometry and NMR applications to simplify spectral interpretation.
  • Available in small pack sizes for research-scale experiments.