Learn More
Medchemexpress LLC 1,2-distearoyl-sn-glycero-3-phosphocholine-d83 | 326495-40-3 | 873.66 g/mol | C44H5D83NO8P | 10 MG
SDP
Catalog No. 5000209327
Encompass_Preferred
DSPC-d83 is the deuterium-labeled form of 1,2-distearoyl-sn-glycero-3-phosphocholine used as a labeled lipid for tracer, internal standard, and lipid formulation research. Supplied as a white to off-white powder for research use only, it is commonly used in liposome and lipid nanoparticle studies and analytical methods such as NMR, GC-MS, and LC-MS.
  • Perdeuterated labeling enables isotopic tracing and quantitative analysis.
  • Suitable for liposome and lipid nanoparticle formulation studies.
  • Provided as a powder for convenient storage and handling.
  • Used as an internal standard for NMR, GC-MS, and LC-MS methods.
  • Intended for research use only; follow the safety data sheet for handling and storage.
Catalog No. 50-002-09327 Supplier Medchemexpress LLC Supplier No. HYW040193S310MG
Add to Cart
Add to Cart
DSPC-d83 is the deuterium-labeled form of 1,2-distearoyl-sn-glycero-3-phosphocholine used as a labeled lipid for tracer, internal standard, and lipid formulation research. Supplied as a white to off-white powder for research use only, it is commonly used in liposome and lipid nanoparticle studies and analytical methods such as NMR, GC-MS, and LC-MS.
  • Perdeuterated labeling enables isotopic tracing and quantitative analysis.
  • Suitable for liposome and lipid nanoparticle formulation studies.
  • Provided as a powder for convenient storage and handling.
  • Used as an internal standard for NMR, GC-MS, and LC-MS methods.
  • Intended for research use only; follow the safety data sheet for handling and storage.