Learn More
Medchemexpress LLC 1,2-distearoyl-sn-glycero-3-phosphoethanolamine conjugated to biotin | 133695-76-8 | MFCD01961045 | 97.5% | 974.36 | C51H96N3O10PS | 50 MG
SDP
Catalog No. 5000280977
Encompass_Preferred
DSPE-Biotin is a biotinylated phospholipid used as a lipid reagent for biochemical research, liposome formulation, and surface functionalization of nanoparticles. It enables biotin-mediated capture or labeling of lipid assemblies and supports drug-delivery and biochemical assay applications. Store cold and protect from light for best stability.
  • Biotinylated phospholipid enabling strong biotin-avidin interactions.
  • Suitable for liposome preparation and nanoparticle surface functionalization.
  • Facilitates biotin-mediated capture and labeling of lipid assemblies.
  • High purity (97.5%) for reproducible experimental results.
  • Solid, off-white to light yellow appearance for easy handling.
  • Storage: -20°C; in solvent: -80°C (6 months), -20°C (1 month); protect from light.
Catalog No. 50-002-80977 Supplier Medchemexpress LLC Supplier No. HYW44100050MG
DSPE-Biotin is a biotinylated phospholipid used as a lipid reagent for biochemical research, liposome formulation, and surface functionalization of nanoparticles. It enables biotin-mediated capture or labeling of lipid assemblies and supports drug-delivery and biochemical assay applications. Store cold and protect from light for best stability.
  • Biotinylated phospholipid enabling strong biotin-avidin interactions.
  • Suitable for liposome preparation and nanoparticle surface functionalization.
  • Facilitates biotin-mediated capture and labeling of lipid assemblies.
  • High purity (97.5%) for reproducible experimental results.
  • Solid, off-white to light yellow appearance for easy handling.
  • Storage: -20°C; in solvent: -80°C (6 months), -20°C (1 month); protect from light.