Learn More
Medchemexpress LLC 1,2-Distearoyl-sn-Glycero-3-Phosphoethanolamine-N-azide | 2839508-98-2 | 96.4% | 831.11 g/mol | C43H83N4O9P | 5 MG
SDP
Catalog No. 5000281100
Encompass_Preferred
DSPE-N3 is an azide-functionalized distearoyl phosphoethanolamine used as a click-chemistry lipid for bioconjugation and nanoparticle formulation. The amphiphilic molecule presents an azide headgroup that enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) to attach alkyne-containing probes, polymers, or targeting ligands, and is commonly applied in liposome and micelle formation, surface functionalization, and targeted delivery research.
  • Contains an azide functional group for bioorthogonal click chemistry
  • Amphiphilic structure supports liposome and micelle formation
  • Suitable for conjugation of alkyne-bearing molecules via CuAAC
  • High chemical purity for research-grade applications
  • Store sealed at 4°C and protect from moisture for stability
  • Available in small lab-scale quantities for formulation work
Catalog No. 50-002-81100 Supplier Medchemexpress LLC Supplier No. HYW4409995MG
DSPE-N3 is an azide-functionalized distearoyl phosphoethanolamine used as a click-chemistry lipid for bioconjugation and nanoparticle formulation. The amphiphilic molecule presents an azide headgroup that enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) to attach alkyne-containing probes, polymers, or targeting ligands, and is commonly applied in liposome and micelle formation, surface functionalization, and targeted delivery research.
  • Contains an azide functional group for bioorthogonal click chemistry
  • Amphiphilic structure supports liposome and micelle formation
  • Suitable for conjugation of alkyne-bearing molecules via CuAAC
  • High chemical purity for research-grade applications
  • Store sealed at 4°C and protect from moisture for stability
  • Available in small lab-scale quantities for formulation work