Learn More
Medchemexpress LLC 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[carboxy(polyethylene glycol)-2000] (sodium salt) | 474922-20-8 | 100.0% | 2000 g/mol (average) | C134H267N2O57P | 5 MG
SDP
Catalog No. 5000284570
Encompass_Preferred
DSPE-PEG-COOH (MW 2000) is a PEGylated phospholipid used to formulate liposomes and lipid nanoparticles for research applications. The DSPE moiety anchors in lipid bilayers while the PEG chain provides steric stabilization; the terminal carboxylic acid enables covalent conjugation to primary amines for surface functionalization.
  • Used to prepare liposomes and lipid nanoparticles for drug delivery research.
  • Terminal carboxylic acid enables amide bond formation with primary amines.
  • PEG2000 chain improves colloidal stability and circulation time.
  • White to off-white solid, suitable for storage and handling in laboratory settings.
  • Available in small pack sizes suitable for research-scale experiments.
Catalog No. 50-002-84570 Supplier Medchemexpress LLC Supplier No. HYW5914615MG
DSPE-PEG-COOH (MW 2000) is a PEGylated phospholipid used to formulate liposomes and lipid nanoparticles for research applications. The DSPE moiety anchors in lipid bilayers while the PEG chain provides steric stabilization; the terminal carboxylic acid enables covalent conjugation to primary amines for surface functionalization.
  • Used to prepare liposomes and lipid nanoparticles for drug delivery research.
  • Terminal carboxylic acid enables amide bond formation with primary amines.
  • PEG2000 chain improves colloidal stability and circulation time.
  • White to off-white solid, suitable for storage and handling in laboratory settings.
  • Available in small pack sizes suitable for research-scale experiments.