Learn More
Medchemexpress LLC 1,2-distearoyl-sn-glycero-3-phosphoethanolamine N-hydroxysuccinimide ester | 1383932-86-2 | MFCD29267127 | 95.0% | 945.21 g/mol | C49H89N2O13P | 25 MG
SDP
Catalog No. 5000281015
Encompass_Preferred
DSPE-NHS is an NHS-activated phospholipid (1,2-distearoyl-sn-glycero-3-phosphoethanolamine N-hydroxysuccinimide ester) used to introduce a reactive NHS ester at liposome or nanoparticle surfaces for covalent conjugation to primary amines on proteins, peptides, and other amine-containing molecules. It self-assembles into lipid bilayers and is commonly used for antibody labeling, liposome functionalization, and nanoparticle surface modification in research applications.
  • Reactive NHS ester for efficient amine coupling.
  • Self-assembles into lipid bilayers for membrane incorporation.
  • Enables stable amide bond formation with proteins and peptides.
  • Suitable for liposome and nanoparticle surface modification.
  • Available in multiple laboratory-scale package sizes for research use.
Catalog No. 50-002-81015 Supplier Medchemexpress LLC Supplier No. HYW44101125MG
DSPE-NHS is an NHS-activated phospholipid (1,2-distearoyl-sn-glycero-3-phosphoethanolamine N-hydroxysuccinimide ester) used to introduce a reactive NHS ester at liposome or nanoparticle surfaces for covalent conjugation to primary amines on proteins, peptides, and other amine-containing molecules. It self-assembles into lipid bilayers and is commonly used for antibody labeling, liposome functionalization, and nanoparticle surface modification in research applications.
  • Reactive NHS ester for efficient amine coupling.
  • Self-assembles into lipid bilayers for membrane incorporation.
  • Enables stable amide bond formation with proteins and peptides.
  • Suitable for liposome and nanoparticle surface modification.
  • Available in multiple laboratory-scale package sizes for research use.