Learn More
Medchemexpress LLC 1,2-Distearoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-2000] sodium | 247925-28-6 | 98.0% | 2808.74 | 50 MG
SDP
Catalog No. 5000435895
Encompass_Preferred
18:0 mPEG2000 PE sodium is a phospholipid-polyethylene glycol conjugate, serving as a key PEG lipid component in lipid nanoparticles (LNPs). It is utilized in gene transfection, drug carrier systems, and drug delivery research. This compound possesses both hydrophilic and hydrophobic properties, enabling it to encapsulate drugs within liposomes, thereby enhancing their stability and efficacy.
  • Conjugate of phospholipid and polyethylene glycol.
  • Serves as a key PEG lipid component in lipid nanoparticles (LNPs).
  • Applicable in gene transfection research.
  • Useful in drug carrier development.
  • Supports drug delivery applications.
  • Exhibits both hydrophilic and hydrophobic characteristics.
  • Capable of encapsulating drugs within liposomes.
  • Enhances drug stability and efficacy.
Catalog No. 50-004-35895 Supplier Medchemexpress LLC Supplier No. HY11276050MG
18:0 mPEG2000 PE sodium is a phospholipid-polyethylene glycol conjugate, serving as a key PEG lipid component in lipid nanoparticles (LNPs). It is utilized in gene transfection, drug carrier systems, and drug delivery research. This compound possesses both hydrophilic and hydrophobic properties, enabling it to encapsulate drugs within liposomes, thereby enhancing their stability and efficacy.
  • Conjugate of phospholipid and polyethylene glycol.
  • Serves as a key PEG lipid component in lipid nanoparticles (LNPs).
  • Applicable in gene transfection research.
  • Useful in drug carrier development.
  • Supports drug delivery applications.
  • Exhibits both hydrophilic and hydrophobic characteristics.
  • Capable of encapsulating drugs within liposomes.
  • Enhances drug stability and efficacy.