Learn More
Medchemexpress LLC 1,2-distearoyl-sn-glycero-3-phosphoethanolamine N-[methoxy(polyethylene glycol)-2000] sodium | 247925-28-6 | MFCD00240077 | 98.0% | 2808.74 g/mol | C45H87NNaO11P | 5 MG
SDP
Catalog No. 5000262299
Encompass_Preferred
18:0 mPEG2000 PE sodium is a PEGylated phosphoethanolamine lipid used as a PEG-lipid component in lipid nanoparticles, micelles, and other drug-delivery formulations to confer steric stabilization and modify circulation properties. Supplied as the sodium salt in a solid form, it is intended for research use in formulation and delivery studies.
  • Provides steric stabilization to nanoparticle surfaces.
  • Suitable for lipid nanoparticle and micelle formulations.
  • Sodium salt form for aqueous compatibility.
  • High purity suitable for research applications.
  • White to off-white solid, easy to handle and store.
  • Soluble in DMSO with warming and sonication.
Catalog No. 50-002-62299 Supplier Medchemexpress LLC Supplier No. HY1127605MG
18:0 mPEG2000 PE sodium is a PEGylated phosphoethanolamine lipid used as a PEG-lipid component in lipid nanoparticles, micelles, and other drug-delivery formulations to confer steric stabilization and modify circulation properties. Supplied as the sodium salt in a solid form, it is intended for research use in formulation and delivery studies.
  • Provides steric stabilization to nanoparticle surfaces.
  • Suitable for lipid nanoparticle and micelle formulations.
  • Sodium salt form for aqueous compatibility.
  • High purity suitable for research applications.
  • White to off-white solid, easy to handle and store.
  • Soluble in DMSO with warming and sonication.