Learn More
Medchemexpress LLC 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[PDP(PEG)-2000] (ammonium salt) | 474922-24-2 | >99.0% | C140H273N4O55PS2 | 10MG
SDP
Catalog No. 5000259154
Encompass_Preferred
DSPE-PEG(2000) PDP is an orthopyridyl-disulfide functionalized PEGylated phospholipid (ammonium salt) used to introduce a reducible disulfide linker for bioconjugation and surface functionalization of liposomes and nanoparticles. It is supplied as a purified solid intended for cold storage and long-term stability.
  • Orthopyridyl-disulfide functionality enables thiol-reactive conjugation.
  • PEG2000 spacer provides steric stabilization and surface shielding.
  • Suitable for liposome and nanoparticle surface modification.
  • High purity supports reliable and reproducible conjugation chemistry.
  • Supplied as a stable solid for cold storage and handling.
  • Enables formation of reducible disulfide linkers for controlled release strategies.
Catalog No. 50-002-59154 Supplier Medchemexpress LLC Supplier No. HY144000A10MG
DSPE-PEG(2000) PDP is an orthopyridyl-disulfide functionalized PEGylated phospholipid (ammonium salt) used to introduce a reducible disulfide linker for bioconjugation and surface functionalization of liposomes and nanoparticles. It is supplied as a purified solid intended for cold storage and long-term stability.
  • Orthopyridyl-disulfide functionality enables thiol-reactive conjugation.
  • PEG2000 spacer provides steric stabilization and surface shielding.
  • Suitable for liposome and nanoparticle surface modification.
  • High purity supports reliable and reproducible conjugation chemistry.
  • Supplied as a stable solid for cold storage and handling.
  • Enables formation of reducible disulfide linkers for controlled release strategies.