Learn More
Medchemexpress LLC 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[poly(ethylene glycol)-folate] | 95.7% | 5000 (average) | 25 MG
SDP
Catalog No. 5000281640
Encompass_Preferred
DSPE-PEG-Folate (MW 5000) is a pegylated lipid conjugate in which a DSPE lipid anchor is linked to a poly(ethylene glycol) chain terminated with folic acid. It is supplied as a solid and is formulated for use in liposomes, micelles, and other nanoparticle systems to enable folate receptor-mediated targeting and uptake.
  • Provides folate receptor targeting for enhanced cellular uptake.
  • Contains a DSPE lipid anchor for stable membrane insertion.
  • Uses a PEG 5000 chain to improve circulation and reduce nonspecific clearance.
  • High purity supports reproducible nanoparticle formulation.
  • Supplied as a solid for convenient storage and handling.
Catalog No. 50-002-81640 Supplier Medchemexpress LLC Supplier No. HYW44089025MG
DSPE-PEG-Folate (MW 5000) is a pegylated lipid conjugate in which a DSPE lipid anchor is linked to a poly(ethylene glycol) chain terminated with folic acid. It is supplied as a solid and is formulated for use in liposomes, micelles, and other nanoparticle systems to enable folate receptor-mediated targeting and uptake.
  • Provides folate receptor targeting for enhanced cellular uptake.
  • Contains a DSPE lipid anchor for stable membrane insertion.
  • Uses a PEG 5000 chain to improve circulation and reduce nonspecific clearance.
  • High purity supports reproducible nanoparticle formulation.
  • Supplied as a solid for convenient storage and handling.