Learn More
Medchemexpress LLC 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-NHS (DSPE-NHS) | 1383932-86-2 | MFCD34561791 | 95.0% | 945.2 g/mol | C49H89N2O13P | 10 MG
SDP
Catalog No. 5000280971
Encompass_Preferred
DSPE-NHS is an N-hydroxysuccinimide (NHS) ester functionalized distearoyl phosphoethanolamine used for bioconjugation and liposome formation. The reagent self-assembles into lipid bilayers in aqueous solution, and the NHS ester reacts selectively with primary amines to form stable amide bonds, enabling antibody labeling and attachment of amine-containing molecules for nanoparticle and drug-delivery applications.
  • Reactive NHS ester for covalent coupling to primary amines.
  • Self-assembles into lipid bilayers in aqueous solution.
  • Enables antibody labeling and protein conjugation.
  • Suitable for liposome and nanocarrier preparation.
  • Long-term storage recommended at -20 °C, sealed and protected from moisture and light.
Catalog No. 50-002-80971 Supplier Medchemexpress LLC Supplier No. HYW44101110MG
DSPE-NHS is an N-hydroxysuccinimide (NHS) ester functionalized distearoyl phosphoethanolamine used for bioconjugation and liposome formation. The reagent self-assembles into lipid bilayers in aqueous solution, and the NHS ester reacts selectively with primary amines to form stable amide bonds, enabling antibody labeling and attachment of amine-containing molecules for nanoparticle and drug-delivery applications.
  • Reactive NHS ester for covalent coupling to primary amines.
  • Self-assembles into lipid bilayers in aqueous solution.
  • Enables antibody labeling and protein conjugation.
  • Suitable for liposome and nanocarrier preparation.
  • Long-term storage recommended at -20 °C, sealed and protected from moisture and light.