Learn More
Medchemexpress LLC 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-PEG-thiol | >95.0% | 25 MG
SDP
Catalog No. 5000280671
Encompass_Preferred
DSPE-PEG-SH (average MW 1,000 Da) is a thiol-terminated PEGylated phospholipid combining a DSPE lipid anchor with a polyethylene glycol spacer that ends in a reactive sulfhydryl. Supplied as a solid, it is used to introduce surface-accessible thiol groups into liposomes, lipid nanoparticles, and other lipid assemblies for bioconjugation and targeted delivery applications.
  • Provides a lipid anchor for stable incorporation into bilayers.
  • Contains a terminal thiol (-SH) for conjugation to thiol-reactive reagents.
  • Average molecular weight ~1,000 Da for the PEG spacer.
  • Facilitates surface functionalization of nanoparticles and liposomes.
  • Typically supplied as a powder with high purity (≥95%).
  • Useful for preparing targeted delivery systems and bioconjugates.
Catalog No. 50-002-80671 Supplier Medchemexpress LLC Supplier No. HYW44089525MG
DSPE-PEG-SH (average MW 1,000 Da) is a thiol-terminated PEGylated phospholipid combining a DSPE lipid anchor with a polyethylene glycol spacer that ends in a reactive sulfhydryl. Supplied as a solid, it is used to introduce surface-accessible thiol groups into liposomes, lipid nanoparticles, and other lipid assemblies for bioconjugation and targeted delivery applications.
  • Provides a lipid anchor for stable incorporation into bilayers.
  • Contains a terminal thiol (-SH) for conjugation to thiol-reactive reagents.
  • Average molecular weight ~1,000 Da for the PEG spacer.
  • Facilitates surface functionalization of nanoparticles and liposomes.
  • Typically supplied as a powder with high purity (≥95%).
  • Useful for preparing targeted delivery systems and bioconjugates.