Learn More
Medchemexpress LLC 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-PEG3-azide | 97.0% | 977.30 | C50H97N4O12P | 10 MG
SDP
Catalog No. 5000298431
Encompass_Preferred
DSPE-PEG3-azide is an azide-functionalized lipid-polyethylene glycol conjugate with a triethylene glycol spacer terminating in an azide. It is provided for research use as a click-chemistry reagent to introduce azide groups into lipid assemblies, enabling surface modification and bioconjugation of micelles and nanoparticle formulations. Certificate of analysis reports a molecular weight of 977.30 and purity of approximately 97% (HPLC).
  • Azide-terminated PEG spacer enables copper-catalyzed or strain-promoted click reactions.
  • Amphiphilic lipid-PEG structure facilitates incorporation into micelles and liposomes.
  • High purity suitable for research bioconjugation and nanoparticle preparation.
  • Available in small pack sizes for method development and scale-up.
  • Stable under typical storage conditions when kept desiccated and cold.
Catalog No. 50-002-98431 Supplier Medchemexpress LLC Supplier No. HY17494510MG
DSPE-PEG3-azide is an azide-functionalized lipid-polyethylene glycol conjugate with a triethylene glycol spacer terminating in an azide. It is provided for research use as a click-chemistry reagent to introduce azide groups into lipid assemblies, enabling surface modification and bioconjugation of micelles and nanoparticle formulations. Certificate of analysis reports a molecular weight of 977.30 and purity of approximately 97% (HPLC).
  • Azide-terminated PEG spacer enables copper-catalyzed or strain-promoted click reactions.
  • Amphiphilic lipid-PEG structure facilitates incorporation into micelles and liposomes.
  • High purity suitable for research bioconjugation and nanoparticle preparation.
  • Available in small pack sizes for method development and scale-up.
  • Stable under typical storage conditions when kept desiccated and cold.