Learn More
Medchemexpress LLC 1,2-distearoyl-sn-glycero-3-phosphorylcholine | 816-94-4 | 100.0% | 790.15 g/mol | C44H88NO8P | 5 G
SDP
Catalog No. 5000208770
Encompass_Preferred
DSPC (1,2-distearoyl-sn-glycero-3-phosphorylcholine) is a cylindrical-shaped phospholipid used to form liposomes and as a structural lipid in lipid nanoparticle (LNP) systems. It is supplied for research use and is commonly employed in membrane model studies and formulation development.
  • Cylindrical phospholipid suitable for liposome and LNP formulations.
  • High purity (99.95%).
  • Molecular formula C44H88NO8P; molecular weight 790.15 g/mol.
  • Common pack sizes include 100 mg, 500 mg, 1 g, 5 g, 10 g, and 50 g.
  • For research use only; not for use in patients.
Catalog No. 50-002-08770 Supplier Medchemexpress LLC Supplier No. HYW0401935G
DSPC (1,2-distearoyl-sn-glycero-3-phosphorylcholine) is a cylindrical-shaped phospholipid used to form liposomes and as a structural lipid in lipid nanoparticle (LNP) systems. It is supplied for research use and is commonly employed in membrane model studies and formulation development.
  • Cylindrical phospholipid suitable for liposome and LNP formulations.
  • High purity (99.95%).
  • Molecular formula C44H88NO8P; molecular weight 790.15 g/mol.
  • Common pack sizes include 100 mg, 500 mg, 1 g, 5 g, 10 g, and 50 g.
  • For research use only; not for use in patients.