Learn More
Medchemexpress LLC 1,2-distearoyl-sn-glycero-3-phosphorylcholine | 816-94-4 | MFCD00036905 | 100.0% | 790.15 | C44H88NO8P | 1 G
SDP
Catalog No. 5000209067
Encompass_Preferred
1,2-distearoyl-sn-glycero-3-phosphorylcholine (DSPC) is a saturated phospholipid used to formulate liposomes and as a structural lipid component in lipid nanoparticle systems. It is a white to off-white solid with high reported purity and defined storage recommendations.
  • Used to synthesize liposomes and as a lipid component in lipid nanoparticle systems.
  • White to off-white solid with high purity.
  • Available in multiple pack sizes, including 1 g.
  • Powder storage: -20°C for long-term stability.
  • In solvent storage: -80°C for up to 6 months, -20°C for short-term storage.
Catalog No. 50-002-09067 Supplier Medchemexpress LLC Supplier No. HYW0401931G
1,2-distearoyl-sn-glycero-3-phosphorylcholine (DSPC) is a saturated phospholipid used to formulate liposomes and as a structural lipid component in lipid nanoparticle systems. It is a white to off-white solid with high reported purity and defined storage recommendations.
  • Used to synthesize liposomes and as a lipid component in lipid nanoparticle systems.
  • White to off-white solid with high purity.
  • Available in multiple pack sizes, including 1 g.
  • Powder storage: -20°C for long-term stability.
  • In solvent storage: -80°C for up to 6 months, -20°C for short-term storage.